Learn More
ATCC Staphylococcus epidermidis; Strain: BPH0695
Catalog No. 502382404
Encompass
Staphylococcus epidermidis strain BPH0695 is a multidrug-resistant bacterium that was isolated in 2007 from human blood in Australia. This strain is glycopeptide heteroresistant and the depositor states that this phenotype is due to RpoB D471E + I527M substitutions.
Catalog No. 50-238-2404 Supplier ATCC Supplier No. BAA3173
Staphylococcus epidermidis strain BPH0695 is a multidrug-resistant bacterium that was isolated in 2007 from human blood in Australia. This strain is glycopeptide heteroresistant and the depositor states that this phenotype is due to RpoB D471E + I527M substitutions.