Learn More
ATCC Staphylococcus epidermidis; Strain PCI 1200
Supplier:  ATCC 12228
Encompass
Staphylococcus epidermidis FDA strain PCI 1200 is a whole-genome sequence bacterial strain that was deposited by FDA. The culture has applications in bioinformatics, food testing, media testing, and quality control, and it is a microbiome standard component.
Catalog No. 50-238-2313
 Restricted Item