Learn More
ATCC Staphylococcus haemolyticus; Strain SM 131
Supplier:  ATCC 29970
Encompass
Staphylococcus haemolyticus strain SM 131 [DSM 20263] is a whole-genome sequenced type strain that was isolated from human skin. This bacterial culture can be used as a quality control strain for API products.
Catalog No. 50-238-3560