Learn More
ATCC Staphylococcus saprophyticus; genomic DNA
Catalog No. 502384619
Encompass
Genomic DNA isolated from Staphylococcus saprophyticus subsp. Saprophyticus (ATCC 15305). This whole-genome sequenced product can be used in PCR and other molecular biology applications.
Catalog No. 50-238-4619 Supplier ATCC Supplier No. 15305D5
Genomic DNA isolated from Staphylococcus saprophyticus subsp. Saprophyticus (ATCC 15305). This whole-genome sequenced product can be used in PCR and other molecular biology applications.