Learn More
ATCC Staphylococcus saprophyticus; subsp. saprophyticus
Catalog No. 502384005
Encompass
Staphylococcus saprophyticus subsp. saprophyticus strain NCTC 7292 [ATCC 19701, NCIB 8711, S-41] is a whole genome-sequenced type strain that was isolated from urine. This bacterium has applications in bioinformatics and quality control.
Catalog No. 50-238-4005 Supplier ATCC Supplier No. 15305
Staphylococcus saprophyticus subsp. saprophyticus strain NCTC 7292 [ATCC 19701, NCIB 8711, S-41] is a whole genome-sequenced type strain that was isolated from urine. This bacterium has applications in bioinformatics and quality control.