Learn More
ATCC Flavonifractor plautii
Supplier:  ATCC 49531
Encompass
This strain was deposited as the type strain for Clostridium orbiscindens. It degrades kaempferol, naringenin, and quercetin.
Catalog No. 50-237-9654