Learn More
Genscript Corporation Staphylokinase 
Supplier:  Genscript Corporation Z027971
Encompass
Staphylokinase (SAK), a 16kDa profibrinolytic protein from the Staphylococcus aureus, has been demonstrated to induce highly fibrin-specific thrombolysis in both human plasma and in limited clinical trials. Recent studies on the thrombolytic potential of recombinant SAK in achieving early perfusion in myocardial infarction and in the dissolution of platelet-rich clot have clearly established its immense utility in clinical medicine as a thrombolytic agent and suggested that it can be developed as a potent clot-dissolving agent.
Catalog No. 50-276-580