Learn More
Med Vet International 9 Lives Autoclavable Surgical Skin Stapler Cartridge of 35 Staples Only, Sterile, J1501F, 5/Pack
Catalog No. 5000495527
Encompass_Preferred
• now priced competitive against suture • simple technique with good cosmetic results Features: •  Fixed angled head allows easy visualization during closure. •  Simple to reload cartridges; for best results autoclave handle and cartridge separately. •  Can view remaining staples on clear bottom. •  Light weight but durable with comfortable ergonomic grip. •  Handle measures 6.5" L x 3" H x 1/2" W •  Staples are 316 L surgical stainless steel Cartridge measures 2 1/4" L x 3/4" W •  Skin Staple after Closure: 3.6mm high x 6.9mm wide The Challenge • Most skin staplers are considered single use devices and cannot be safely autoclaved. • 35 staples is the standard cartridge size. The average vet surgery uses only 7-10 staples. 9 Lives Autoclavable Surgical Skin Stapler Cartridge of 35 Staples Only, 5 pk, Sterile
Catalog No. 50-004-95527 Supplier Med Vet International Supplier No. JOR1501F
• now priced competitive against suture • simple technique with good cosmetic results Features: •  Fixed angled head allows easy visualization during closure. •  Simple to reload cartridges; for best results autoclave handle and cartridge separately. •  Can view remaining staples on clear bottom. •  Light weight but durable with comfortable ergonomic grip. •  Handle measures 6.5" L x 3" H x 1/2" W •  Staples are 316 L surgical stainless steel Cartridge measures 2 1/4" L x 3/4" W •  Skin Staple after Closure: 3.6mm high x 6.9mm wide The Challenge • Most skin staplers are considered single use devices and cannot be safely autoclaved. • 35 staples is the standard cartridge size. The average vet surgery uses only 7-10 staples. 9 Lives Autoclavable Surgical Skin Stapler Cartridge of 35 Staples Only, 5 pk, Sterile