Learn More
Electron Microscopy Sciences Star Test Chart Sector Pitch 10 R65
SDP
Catalog No. 501901826
Encompass_Preferred
Siemens Star or Sunburst Pattern vacuum deposited chrome image. Outside diameter of image 25 mm. Normal substrate B270 Glass. Overall size 50 mm x 50 mm. With 36 Sectors and 10 degree pitch. Unresolved centre core 0.2mm.
Catalog No. 50-190-1826 Supplier Electron Microscopy Sciences Supplier No. 6809808
Siemens Star or Sunburst Pattern vacuum deposited chrome image. Outside diameter of image 25 mm. Normal substrate B270 Glass. Overall size 50 mm x 50 mm. With 36 Sectors and 10 degree pitch. Unresolved centre core 0.2mm.