Learn More
Electron Microscopy Sciences Star Test Chart Sector Pitch 5
SDP
Catalog No. 501901578
Encompass_Preferred
Siemens Star or Sunburst Pattern vacuum deposited chrome image. Outside diameter of image 25 mm. Normal substrate B270 Glass. Overall size 50 mm x 50 mm. With 72 Sectors and 5 degree pitch. Unresolved centre core 0.4mm.
Catalog No. 50-190-1578 Supplier Electron Microscopy Sciences Supplier No. 6809809
Siemens Star or Sunburst Pattern vacuum deposited chrome image. Outside diameter of image 25 mm. Normal substrate B270 Glass. Overall size 50 mm x 50 mm. With 72 Sectors and 5 degree pitch. Unresolved centre core 0.4mm.