Learn More
World Precision Instrument Starbismus scissors, 12 cm, Curved, Black, Round Tips
SDP
Supplier:  World Precision Instrument WPB409000
Encompass
Strabismus scissors, available in straight and curved varieties, are vital tools in ophthalmic surgery, particularly in the correction of strabismus or ocular misalignment. The straight design of these scissors, facilitates precise cutting in targeted areas during strabismus correction, offering you accuracy and control. Conversely, curved strabismus scissors feature blades with a gentle curve, enhancing access to specific areas and allowing controlled cutting around curves and corners, making them ideal for nuanced and intricate surgical maneuvers. Both types are coated in black, ensuring durability by resisting corrosion and minimizing glare under surgical lights for improved visibility. These scissors play a crucial role in various ophthalmic procedures, contributing to the alignment of eyes and precise cutting around delicate eye structures. They are made of stainless steel and may be sterilized using an autoclave or a chemical sterilant.
Catalog No. 50-253-8269