Learn More
Alkali Scientific Starch/Casein Agar 20mL Plate , 10/Pk
SDP
Catalog No. 5000662832
Encompass_Preferred
Starch Casein Agar is a specialized culture medium used for isolating and studying microorganisms capable of hydrolyzing complex carbohydrates and proteins. The medium contains starch and casein, which serve as substrates for enzymatic activity. Microorganisms that produce amylase enzymes break down starch surrounding their colonies, while organisms producing proteases degrade casein proteins. After incubation, starch hydrolysis can be visualized by adding iodine, which stains intact starch while leaving clear zones around colonies that degraded it. This medium is particularly useful for isolating actinomycetes from soil and environmental samples and for studying microbial enzyme activity.
Catalog No. 50-006-62832 Supplier Alkali Scientific Supplier No. SCSP00
Starch Casein Agar is a specialized culture medium used for isolating and studying microorganisms capable of hydrolyzing complex carbohydrates and proteins. The medium contains starch and casein, which serve as substrates for enzymatic activity. Microorganisms that produce amylase enzymes break down starch surrounding their colonies, while organisms producing proteases degrade casein proteins. After incubation, starch hydrolysis can be visualized by adding iodine, which stains intact starch while leaving clear zones around colonies that degraded it. This medium is particularly useful for isolating actinomycetes from soil and environmental samples and for studying microbial enzyme activity.