Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Starch, Corn, Reagents

Catalog No. 01336944
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-944 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-336-944 Supplier Reagents Holdings Llc Supplier No. C2331000500A5
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Starch, Corn, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

CAS 9005-25-8
Boiling Range 668°C
Color White
Boiling Point 668°C
Packaging Poly Bottle
Molecular Formula C12H22O11
Quantity 500 g
Specific Gravity 1.5
InChI Key GUBGYTABKSRVRQ-UHFFFAOYNA-N
SMILES OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O
IUPAC Name 2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
Molecular Weight (g/mol) 342.30
Grade Reagent
Physical Form Solid
Chemical Name or Material Starch, Corn
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.