Learn More
Sigma Aldrich Fine Chemicals Biosciences Starch from corn practical1KG
Catalog No. 502899895
Encompass
Starch is a natural storage carbohydrate polymer produced by plants and varies in its form and function. It is considered as a macroconstituent of food and serves as an energy source. It is renewable and can be biodegraded. Glucose polymers amylose and amylopectin forms starch molecule. Starch influences the food structure and quality. It is a native high amylose corn starch.
Catalog No. 50-289-9895 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. S41801KG
Starch is a natural storage carbohydrate polymer produced by plants and varies in its form and function. It is considered as a macroconstituent of food and serves as an energy source. It is renewable and can be biodegraded. Glucose polymers amylose and amylopectin forms starch molecule. Starch influences the food structure and quality. It is a native high amylose corn starch.