Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Starch (from potato) | 9005-25-8 | 100 G
SDP

Catalog No. 5000411004
Encompass_Preferred

Starch from potato is a biochemical reagent suitable for life science related research, functioning as a biological material or organic compound. It is intended for research use only.

  • Appearance: Solid
  • Color: White to off-white
  • Formula: (C6H10O5)n
  • Shipping at room temperature in continental US
  • Storage: Powder at -20°C for 3 years or 4°C for 2 years; in solvent at -80°C for 6 months or -20°C for 1 month
  • Solubility in DMSO: 12.5 mg/mL

Catalog No. 50-004-11004 Supplier Medchemexpress LLC Supplier No. HYB2225A100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Starch from potato is a biochemical reagent suitable for life science related research, functioning as a biological material or organic compound. It is intended for research use only.

  • Appearance: Solid
  • Color: White to off-white
  • Formula: (C6H10O5)n
  • Shipping at room temperature in continental US
  • Storage: Powder at -20°C for 3 years or 4°C for 2 years; in solvent at -80°C for 6 months or -20°C for 1 month
  • Solubility in DMSO: 12.5 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.