Learn More
Starch (GO/P) Assay Kit, MilliporeSigma™ Supelco™
Catalog No. 111002711 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
11-100-2711 1 Kit
1 options
Catalog No. 11-100-2711 Supplier MilliporeSigma Supelco Supplier No. STA201KT
Add to Cart
Add to Cart
Starch (GO/P) Assay Kit was used to determine the starch content, during an experimental study done in order to investigate the composition and structure of sorghum spent grains (SSG), which is a brewery co-product and is reported to have potential applications thereby adding value to the product. It was also used in starch extraction and quantification, during investigating the importance of OsGA2ox5 in GAs homeostasis, development, gravity responses and stress tolerance in rice. Furthermore, it also found application in being used in analysing factors affecting polyhydroxybutyrate accumulation , which are basically linear biodegradable polyesters produced by bacteria as a carbon store, in mesophyll cells of sugarcane and switchgrass.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Kit
Concentration or Composition (by Analyte or Components) alpha-Amylase, Heat Stable 0.5 mL; Starch Assay Reagent 1 mL/vial; Glucose Oxidase/Peroxidase Reagent 1 capsule; o-Dianisidine Reagent 5 mg; Glucose Standard Solution 0.5 mL; Wheat Starch 1 g; Corn Starch 1 g