Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Starch Ind. 0.5%
SCI_ED

Catalog No. s25581 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
0.5% Solution
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25581 500 mL 0.5% Solution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25581
Only null left
Add to Cart
Add to Cart

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 9005-25-8
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00082026,MFCD00132834
InChI Key GUBGYTABKSRVRQ-UHFFFAOYNA-N
Synonym alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose
PubChem CID 439341
ChEBI CHEBI:18167
IUPAC Name 2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
SMILES OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O

Specifications

Assay Percent Range 0.5% w/v
Density 1.5g/mL
Quantity 500 mL
Packaging 0.5% Solution
Chemical Name or Material Starch
Safety handling Check Alert storage code Red
ShelfLife 2 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.