Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Starch Indicator, 0.5% (w/v), Mercury Free, for Iodometric Titrations, Ricca Chemical
SDP

Catalog No. 800016 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
8000-16 500 mL Natural Poly Bottle
R8000000100 100 mL Natural Poly Bottle
8000-32 1 L Natural Poly Bottle
R80000004F 4 L Cubitainer
8000-1 4 L Natural Poly Bottle
8000-5 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 8000-16 Supplier Ricca Chemical Company Supplier No. 800016
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Iodometric Titrations

Specifications

Chemical Name or Material Starch Indicator
CAS 9005-84-9, 69-72-7, 7732-18-5
Concentration 0.5% (w/v)
Percent Purity 0.5% W/V
Grade Indicator
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Color White
pH 5 to 7
Quantity 500 mL
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.