Learn More
Starch Indicator, 0.5% (w/v), Mercury Free, for Iodometric Titrations, Ricca Chemical
Shop All Ricca Chemical Company Products

Specifications
Specifications
| Chemical Name or Material | Starch Indicator |
| CAS | 9005-84-9, 69-72-7, 7732-18-5 |
| Concentration | 0.5% (w/v) |
| Packaging | Natural Poly Bottle |
| Solvent or Matrix | Aqueous Solution |
| Color | White |
| pH | 5 to 7 |
| Quantity | 1 L |
| Physical Form | Liquid |
| Percent Purity | 0.5% W/V |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.