Learn More
Starch Indicator, 0.5% w/v, Reagents
Specifications
Specifications
| Chemical Name or Material | Starch Indicator, 0.5% w/v |
| CAS | 7732-18-5, 9005-84-9, 69-72-7 |
| Boiling Point | 100°C |
| Percent Purity | 0.5% |
| Grade | Reagent |
| Packaging | Poly Bottle |
| Specific Gravity | 1.0021 |
| Color | White |
| Quantity | 500 mL |
| Physical Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.