Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thomas Scientific Starch Indicator 1% (w/v) 4L
SDP

Catalog No. 5000550977
Encompass_Preferred

Starch Indicator 1% (w/v) 4L

Catalog No. 50-005-50977 Supplier Thomas Scientific Supplier No. C748C73
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Starch Indicator 1% (w/v) 4L

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.