Learn More
Aqua Solutions STARCH INDICATOR .5% 4L
SDP
Catalog No. NC9893731
Encompass_Preferred
Starch Indicator Solution; 0.5% w/v, APHA Indicator Solution; Clear to slightly turbid solution, odorless; 4L; M.P. ~0 deg. C; B.P. ~100 deg. C; Specific Gravity ~1; Percent Volatile by Volume:> 99%; Stored in a cool dry place
Catalog No. NC9893731 Supplier Aqua Solutions Supplier No. 88604LC
Starch Indicator Solution; 0.5% w/v, APHA Indicator Solution; Clear to slightly turbid solution, odorless; 4L; M.P. ~0 deg. C; B.P. ~100 deg. C; Specific Gravity ~1; Percent Volatile by Volume:> 99%; Stored in a cool dry place