Learn More
Starch Indicator, Preserved with Chloroform (Mercury Free), Certified, 1%, LabChem™
Shop All LabChem ProductsSpecifications
Specifications
| Chemical Name or Material | Starch Indicator |
| Name Note | Preserved with Chloroform (Mercury Free) |
| CAS Max % | 98.5, 1, 0.495, 0.005 |
| CAS | 9005-25-8, 67-66-3, 64-17-5 |
| Identification | Passes Test |
| Packaging | Poly Bottle |
| Specific Gravity | 1g/mL |
| Suitability | Passes Test |
| Decomposition Information | Hydrogen chloride; Carbon monoxide; Carbon dioxide |
| Density | 1g/mL |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.