Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Starch Indicator, Preserved with Chloroform (Mercury Free), Certified, 1%, LabChem™

Catalog No. LC253154 Shop All LabChem Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC253154 4 L
LC253151 500 mL
LC253152 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC253154 Supplier LabChem, Inc. Supplier No. LC253154
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Starch Indicator
Name Note Preserved with Chloroform (Mercury Free)
CAS Max % 98.5, 1, 0.495, 0.005
CAS 9005-25-8, 67-66-3, 64-17-5
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1g/mL
Suitability Passes Test
Decomposition Information Hydrogen chloride; Carbon monoxide; Carbon dioxide
Density 1g/mL
Color Colorless
Solubility Information Soluble in water
Quantity 4 L
Physical Form Liquid
Percent Purity 1%
Grade Certified
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.