Learn More
Aqua Solutions Starch Indicator Solution, 1 L
SDP
Catalog No. NC1446738
Encompass
Oxidation-Reduction Indicator for iodometric titrations.
Catalog No. NC1446738 Supplier Aqua Solutions Supplier No. HA34953
Oxidation-Reduction Indicator for iodometric titrations.