Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Starch Indicator Solution; Lovibond™

Catalog No. 09534711 Shop All Tintometer Group Products
Change view
Click to view available options
Quantity:
65 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-534-711 65 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-534-711 Supplier Tintometer Group Supplier No. R50092
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Starch Indicator Solution
CAS 69-72-7, 9005-84-9
Color Colorless
Quantity 65 mL
Physical Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.