Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Starch - Iodide Solution, 0.5% (w/v), Ricca Chemical
SDP

Catalog No. R8056500120 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R8056500120 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R8056500120 Supplier Ricca Chemical Company Supplier No. R8056500120B
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Starch - Iodide Solution
Color White
CAS 7732-18-5, 7681-11-0, 9005-84-9
CAS Min % 97.0, 0.49, 0.49
CAS Max % 100.0, 0.51, 0.51
Percent Purity 0.5%
Grade Laboratory
Concentration or Composition (by Analyte or Components) 0.5% (w/v) Aqueous
Odor Odorless
Packaging Amber Poly Bottle
Quantity 120 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.