Learn More
Starch Solution, 1% w/v Mercury-Free, Reagents
Specifications
Specifications
| Chemical Name or Material | Starch Solution, 1% w/v Mercury-Free |
| Boiling Point | 100°C |
| CAS | 7732-18-5, 9005-84-9, 9005-84-9 |
| Packaging | Poly Bottle |
| Specific Gravity | 1.01 |
| Color | White |
| Quantity | 1 L |
| Physical Form | Liquid |
| Percent Purity | 1% |
| Grade | Reagent |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.