Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Starch Solution, 2% w/v, Mercury-Free, with Salicylic Acid Preservative, Reagents

Catalog No. 01334046
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-334-046 500 mL
01-334-049 1 L
01-334-047 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-334-046 Supplier Reagents Holdings Llc Supplier No. CS001600500A
Only null left
Add to Cart
Add to Cart

Starch Solution, 2% w/v, Mercury-Free, with Salicylic Acid Preservative, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Starch Solution, 2% w/v, Mercury-Free, with Salicylic Acid Preservative
Boiling Point 100°C
CAS 7732-18-5, 9005-84-9, 69-72-7
Percent Purity 2%
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.02
Color White
Quantity 500 mL
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.