Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Starplex™ Scientific StarCLICK 100 Specimen Container

Catalog No. 22046575 Shop All Starplex Scientific Products
Change view
Click to view available options
Closure Color:
Orange
Yellow
Label Description:
No label
Patient label
Sterility:
Non-sterile
Sterile
No. per Pack:
100 Pk.
400 Pk.
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Color Label Description Sterility No. per Pack
22-046-575 Orange Patient label Sterile 100 Pk.
22-045-690 Yellow Patient label Non-sterile 100 Pk.
22-045-680 Orange No label Sterile 100 Pk.
22-046-576 Yellow Patient label Non-sterile 400 Pk.
22-046-578 Yellow No label Non-sterile 400 Pk.
22-046-579 Orange Patient label Sterile 400 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 22-046-575 Supplier Starplex™ Scientific Supplier No. B1002OCC
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

StarCLICK 100 features an audible, double click to confirm the cap is applied correctly. Ideal for leakproof specimen transport.

  • Polypropylene
  • Leakproof transport
  • Clear graduations on two sides in ounces and milliliters
  • Wide mouth design
  • Stackable
  • Available in sterile and non-sterile format
  • Externally validated to meet 95 kPa and IATA standards

Specifications

Closure Color Orange
Disposable Single-use
Material Polypropylene
Sterility Sterile
Closure Material Polyethylene
Description Specimen container
For Use With (Application) Specimen collection and transport
Label Description Patient label
Latex Free Yes
No. per Case 400/Cs.
No. per Pack 100 Pk.
Storage Requirements Ambient
Volume (English) 3 oz
Volume (Metric) 100 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.