Learn More
Quantimetrix QuanTtest Protein Standards, 4 Level Set
SDP
Catalog No. NC9876145
Encompass_Preferred
• For quantitative determination of protein in cerebrospinal fluid and urine for both manual and automated systems • Standards and Controls are stable until the expiration date when stored at 2ºC–8ºC • The kit consists of four protein standards:Level 2 (25 mg/dL), Level 3 (50 mg/dL), Level 4 (100 mg/dL), and Level 5 (200 mg/dL)
Catalog No. NC9876145 Supplier Quantimetrix Supplier No. 341002
• For quantitative determination of protein in cerebrospinal fluid and urine for both manual and automated systems • Standards and Controls are stable until the expiration date when stored at 2ºC–8ºC • The kit consists of four protein standards:Level 2 (25 mg/dL), Level 3 (50 mg/dL), Level 4 (100 mg/dL), and Level 5 (200 mg/dL)