Learn More
Aviva Systems Biology Corp STARD6 Antibody - N-terminal region (ARP65716_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65716_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against STARD6. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N-terminal region of Human STARD6. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-288-550