Learn More
Aviva Systems Biology Corp STARD8 Antibody - N-terminal region (ARP59662_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59662_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against STARD8. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human STARD8. Alternate names: DKFZp686H1668, DLC3, KIAA0189, STARTGAP3, ARHGAP38ApplicationWB. Species reactivity: Dog, Human
Catalog No. 50-286-771