Learn More
MESA LABS BOZEMAN MFG FACILITY SterilAmp II & 5230 ( 26mm x 6.5mm )
SDP
Catalog No. NC0315154
Encompass
SterilAmp II is a compact, self-contained biological indicator for monitoring industrial steam sterilization of liquids in small containers. SterilAmp II precisely integrates the process lethality. SterilAmp II contains a specially formulated culture medium that turns a dramatic yellow when spores grow. The SterilAmp II provides easy visual confirmation of sterilization ~5 days, without the need for difficult and time-consuming laboratory work. Small, conveniently-shaped ampoule ( 26mm x 6.5mm ). No laboratory required. Dramatic color change
Catalog No. NC0315154 Supplier MESA LABS BOZEMAN MFG FACILITY Supplier No. SASU/6
SterilAmp II is a compact, self-contained biological indicator for monitoring industrial steam sterilization of liquids in small containers. SterilAmp II precisely integrates the process lethality. SterilAmp II contains a specially formulated culture medium that turns a dramatic yellow when spores grow. The SterilAmp II provides easy visual confirmation of sterilization ~5 days, without the need for difficult and time-consuming laboratory work. Small, conveniently-shaped ampoule ( 26mm x 6.5mm ). No laboratory required. Dramatic color change