Learn More
Starna Cells Inc STARNA CUVETTE 16.160F-Q10/Z15
SDP
Catalog No. NC9243338
Encompass_Preferred
Fluorimeter, Sub-micro, Square open top, Spectrosil quartz, 2W x 8H, 12.5L x 12.5W x 45H, 0.160mL, Open top, supplied with non-sealing PTFE cover as well as a vaned lid to provide a liquid-tight seal
Catalog No. NC9243338 Supplier Starna Cells Inc Supplier No. 16.160FQ10/Z15
Fluorimeter, Sub-micro, Square open top, Spectrosil quartz, 2W x 8H, 12.5L x 12.5W x 45H, 0.160mL, Open top, supplied with non-sealing PTFE cover as well as a vaned lid to provide a liquid-tight seal