Learn More
Starna Scientific™ DNACON10X-SC2 Reference Solution 10X
Catalog No. 13400415 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Reference Standard Solution
Catalog No. Type
13-400-415 Reference Standard Solution
1 options
Catalog No. 13-400-415 Supplier Starna Scientific™ Supplier No. 869193700
Add to Cart
Add to Cart
Starna Scientific DNACON10X-SC2 is a A260/A280 reference solution aimed at customers wishing to confirm the performance of their NanoDrop™ microvolume UV-Vis spectrophotometer.
  • Developed for the validation of DNA purity measurements
  • Ideal for qualifying conventional instruments when they are used with ultra-low volume cells
TRUSTED_SUSTAINABILITY
Specifications
Concentration 10X
Type Reference Standard Solution
Quantity 2 x 1.5 mL vials
WARNING: Cancer - www.P65Warnings.ca.gov