Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Starplex™ Scientific Anaerobic Transport System

Catalog No. 1490552 Shop All Starplex Scientific Products
Change view
Click to view available options
Quantity:
10 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-905-52 10 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-905-52 Supplier Starplex™ Scientific Supplier No. S120D
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Tube and two swabs in sterile wrap

Single format for swabs, fluid or tissue

  • Self-contained, sterile, ready-to-use system includes tube containing 7mL pre-reduced, semi-solid medium
  • Media indicator turns lavender if anaerobic conditions are compromised
  • Maintains viability for a wide variety of microorganisms for up to 48 hours
  • System features a screw-on cap with a pierceable rubber center, allowing fluid injection without risking oxygen exposure
  • Scored, plastic swab shaft breaks easily to fit the tube

Specifications

Product Type Anaerobic Transport System
For Use With (Application) Maintains viability for a wide variety of microorganisms for up to 48 Hours
Includes Tube and two swabs in sterile wrap
Quantity 10 Pk.
Shaft Material Plastic
Storage Requirements 4°C to 25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.