Learn More
Starplex™ Scientific Kangapack™ Specimen Containers with O-Ring Cap
Catalog No. p-3360613 Shop All Starplex Scientific Products
Change view
Click to view available options
Capacity (Metric):
20 mL
90 mL
Product Type:
Primary Container
Secondary Container
Closure Type:
DisCap
O-Ring Cap
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Product Type Closure Type
14-905-58 20 mL Secondary Container DisCap
14-905-55 90 mL Primary Container O-Ring Cap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 14-905-58 Supplier Starplex™ Scientific Supplier No. B2021WNL
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Patented design nests secondary container inside the primary container for convenient handling of split specimens
  • Minimum fill line on primary and secondary containers
  • O-ring cap design grips top and sides of container for seal
Specifications
Product Type Secondary Container
Material Plastic
Capacity (Metric) 20 mL
Closure Type DisCap
Color White