Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Starplex™ Scientific Dip N Count™ Dip Paddles

Catalog No. 22045173 Shop All Starplex Scientific Products
Change view
Click to view available options
Media combination: MacConkey and CLED
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
22-045-173 Media combination: MacConkey and CLED
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-045-173 Supplier Starplex Scientific Supplier No. B30U0

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

  • For microbial growth
  • Available in culture media combination: CLED Agar and MacConkey Agar with Crystal Violet

Specifications

Content And Storage 4°C to 25°C
Manufacturing Origin United States
Media Type CLED Agar and MacConkey Agar with Crystal Violet
For Use With (Application) For microbial growth
Packaging Type Slide
Quantity 10 Boxes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.