Learn More
Starplex™ Scientific Slide Holder
Catalog No. 14375433 Shop All Starplex Scientific Products
Change view
Click to view available options
Slide Capacity:
4
Catalog No. Slide Capacity
14-375-433 4
1 options
This item is not returnable. View return policy
Catalog No. 14-375-433 Supplier Starplex™ Scientific Supplier No. V302SH
Add to Cart
Add to Cart
This item is not returnable. View return policy
This unlabeled product is suitable for use as a mailer
Manufactured using medical grade plastic that ensures the consistency and reliability of the product. Slide holders are leakproof and travel well during harsh transport or within a pneumatic tube system.
  • Ideal for transporting up to four standard size slides with or without fixative
  • Slides do not touch and remain protected from breakage within each groove
Specifications
Slide Capacity 4
For Use With (Equipment) Microscope Slide
Includes Groove inserts and red O-ring cap
Material Polypropylene