Learn More
Starplex™ Scientific Specimen Containers: Cups
Catalog No. 09735205 Shop All Starplex Scientific Products
Change view
Click to view available options
Capacity (Metric):
30 mL
90 mL
120 mL
192 mL
Sterility:
Non-sterile
Sterile
Closure Type:
Cap with Paddle
Not Included
Spout Cap
Wide mouth
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Sterility Closure Type
09-735-205 120 mL Non-sterile Not Included
14-375-435 30 mL Sterile Cap with Paddle
22-146-593 90 mL Non-sterile Wide mouth
14-904-54 192 mL Non-sterile Spout Cap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 09-735-205 Supplier Starplex™ Scientific Supplier No. B1202VO
Add to Cart
Add to Cart
This item is not returnable. View return policy
  • Break- and leak-resistant for safety during harsh transport conditions or pneumatic tube systems
  • Clarified polypropylene cups provide easy visual analysis of sample
  • Full-circle thread design prevents cross-threading
  • Graduated markings allow quick measurement of sample
  • Labeling provides ample space for patient information
  • Ridges on cap provide a secure grip for safe handling
Specifications
Closure Included No
Material Polypropylene
Sterility Non-sterile
Capacity (Metric) 120 mL
Closure Type Not Included