Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Starplex™ Scientific Starswab II™ Collection and Transport Systems: Amies Clear Medium

Catalog No. 1490592 Shop All Starplex Scientific Products
Change view
Click to view available options
Media Type:
Liquid Amies
Modified Amies Clear Gel
Shaft Material:
Plastic
Straight Wire
Swab Configuration:
Dual Applicator
Minitip flocked
Single Applicator
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Media Type Shaft Material Swab Configuration
14-905-92 Liquid Amies Plastic Dual Applicator
14-905-80 Modified Amies Clear Gel Plastic Dual Applicator
14-905-91 Liquid Amies Plastic Single Applicator
14-905-67 Modified Amies Clear Gel Plastic Single Applicator
14-905-81 Modified Amies Clear Gel Straight Wire Minitip flocked
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 14-905-92 Supplier Starplex™ Scientific Supplier No. SP130LD
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Protective overskirt on cap

  • Designed for collecting, transporting and maintaining the viability of aerobic, anaerobic and facultative microorganisms
  • Swab tips available in single, double or straight wire format
  • Applicators have color-coded caps with overskirt and biohazard plug-cap rings; extra-long swab shafts; and a labeled, round-bottom, spiral transport tube
  • Transport tubes contain 4mL of semi-solid media
  • Spiral media chamber within a seamless polypropylene tube protects sample and keeps it immersed in media during transport
  • Labels preprinted with lot number, expiration date and product code with space reserved for recording patient information
  • Gamma sterilized and wrapped in sterile, easy-to-peel envelope

Amies Clear Medium

  • General-purpose transport medium for recovery of a wide range of organisms, including certain anaerobes
  • Provides excellent survival characteristics for transport and gram stain compatibility

Specifications

Product Type Collection and Transport System
Media Type Liquid Amies
Swab Configuration Dual Applicator
Closure Color White
For Use With (Application) Collecting, transporting and maintaining the viability of aerobic, anaerobic and facultative microorganisms
Packaging Bag
Quantity 500 Cs.
Shaft Material Plastic
Storage Requirements 4°C to 25°C
Tip Material Rayon
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.