Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Starplex™ Scientific Starswab II™ Collection and Transport Systems: Liquid Stuart's Medium
GSA_VA

Catalog No. 1490585 Shop All Starplex Scientific Products
Change view
Click to view available options
Closure Color:
Black
Orange
Teal
Shaft Material:
Plastic
Straight Wire
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Color Shaft Material
14-905-85 Teal Plastic
14-905-87 Orange Straight Wire
14-905-86 Black Plastic
22-046-425 - -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 14-905-85 Supplier Starplex™ Scientific Supplier No. SP132
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Protective overskirt on cap

  • Designed for collecting, transporting and maintaining the viability of aerobic, anaerobic and facultative microorganisms
  • Swab tips available in single, double or straight wire format
  • Applicators have color-coded caps with overskirt and biohazard plug-cap rings; extra-long swab shafts; and a labeled, round-bottom, spiral transport tube
  • Transport tubes contain 4mL of semi-solid media
  • Spiral media chamber within a seamless polypropylene tube protects sample and keeps it immersed in media during transport
  • Labels preprinted with lot number, expiration date and product code with space reserved for recording patient information
  • Gamma sterilized and wrapped in sterile, easy-to-peel envelope

Liquid Stuart's Medium

  • Provide excellent recovery in routine bacteriology and is compatible with gram stain and many rapid screening methods

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Product Type Collection and Transport System
Media Type Liquid Stuart
Swab Configuration Single Applicator
Certifications/Compliance CE
Closure Color Teal
For Use With (Application) Collecting, transporting and maintaining the viability of aerobic, anaerobic and facultative microorganisms
Length (English) 6 in.
Length (Metric) 15.2 cm
Packaging Bag
Quantity 50 Pk.
Shaft Material Plastic
Storage Requirements 4°C to 25°C
Tip Material Rayon
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.