Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Starplex™ Scientific Starswabs II™ Dry Swabs

Catalog No. 1490550 Shop All Starplex Scientific Products
Change view
Click to view available options
Packaging:
Tube
Tip Material:
Rayon
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Tip Material
14-905-50 Tube Rayon
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-905-50 Supplier Starplex™ Scientific Supplier No. S09D
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dry, sterile swabs with plastic shafts

Specifications

Tip Material Rayon
Shelf Life 2 Years
Sterility Sterile
Packaging Tube
For Use With (Application) For assessing microbial presence on various surfaces and equipment
Product Type Starswabs II™ Dry Swab
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.