Learn More
Starplex™ Scientific Starswabs II™ Clinical Swabs
Catalog No. 22046550 Shop All Starplex Scientific Products
Change view
Click to view available options
Tip Type:
Inert swab tip
Catalog No. Tip Type
22-046-550 Inert swab tip
1 options
This item is not returnable. View return policy
Catalog No. 22-046-550 Supplier Starplex™ Scientific Supplier No. S09
Add to Cart
Add to Cart
This item is not returnable. View return policy
Rayon swab tip is inert, non-toxic and permits sample retrieval and absorption
  • Permits sample retrieval and absorption
  • Color-coded caps with protective overskirt
  • Cap rings ensure containment of biohazardous material
  • Thick tube wall protects medium from dehydration
  • Spiral media chamber keeps sample immersed and protected during transport
  • Round bottom tube for easy racking and seamless moulding ensures tube strength
  • Thick, metalized bag prevents premature medium dehydration and protects media from harmful UV rays
  • Zip lock closure allows bag to be resealed
Specifications
Tip Material Rayon
Tip Type Inert swab tip
Product Type Clinical swab