Learn More
MSC Starrett Dial Depth Gage, Hardened Stainless Steel Base, 0.001 in. Graduation, 1 ea
Catalog No. 5000511672
Encompass
The 450 Dial Depth Gage is a precise measurement tool designed for determining the depth of holes, slots, shoulders, and recesses. Featuring a hardened stainless steel base and a rack-and-gear mechanism, this instrument provides reliable readings with an anti-backlash control system for increased accuracy.
  • Graduation of 0.001 in. with 0.100 in. per revolution
  • Hardened stainless steel base for durability
  • Positive split gear anti-backlash control for consistent performance
  • Lock screw for securing the dial bezel and measuring rod
  • Removable hook attachment enables measurements from the edge of a workpiece to slots or shoulders
  • Optional base extensions of 175 mm (7 in.) and 300 mm (12 in.) available
Catalog No. 50-005-11672 Supplier MSC Supplier No. 86447679
The 450 Dial Depth Gage is a precise measurement tool designed for determining the depth of holes, slots, shoulders, and recesses. Featuring a hardened stainless steel base and a rack-and-gear mechanism, this instrument provides reliable readings with an anti-backlash control system for increased accuracy.
  • Graduation of 0.001 in. with 0.100 in. per revolution
  • Hardened stainless steel base for durability
  • Positive split gear anti-backlash control for consistent performance
  • Lock screw for securing the dial bezel and measuring rod
  • Removable hook attachment enables measurements from the edge of a workpiece to slots or shoulders
  • Optional base extensions of 175 mm (7 in.) and 300 mm (12 in.) available