Learn More
MSC Starrett Adjustable Parallel Set, 3/8 to 2-1/4 in, 9/32 in, 6 Piece
Catalog No. 5000535634
Encompass
This adjustable parallel set is designed for layout, gaging, inspection, and machine tool setups. Its adjustable design allows precise micrometer adjustments, replacing multiple solid parallels. It also functions as a gage for checking slot and opening sizes, suitable for machine vises and leveling work. All parallels are 9/32 in (7 mm) thick and slide smoothly.
  • Adjustable feature for exact micrometer size adjustments.
  • Replaces multiple solid parallels.
  • Useful as gages for checking slot and opening sizes.
  • Convenient for machine vises, leveling, and adjusting work on various machines.
  • Parallels slide smoothly for easy adjustment.
  • All parallels are 9/32 in (7 mm) thick.
  • Smaller sizes lock with one screw, larger sizes with two.
  • Includes a set of 6 parallels in a case.
Catalog No. 50-005-35634 Supplier MSC Supplier No. 86405883
This adjustable parallel set is designed for layout, gaging, inspection, and machine tool setups. Its adjustable design allows precise micrometer adjustments, replacing multiple solid parallels. It also functions as a gage for checking slot and opening sizes, suitable for machine vises and leveling work. All parallels are 9/32 in (7 mm) thick and slide smoothly.
  • Adjustable feature for exact micrometer size adjustments.
  • Replaces multiple solid parallels.
  • Useful as gages for checking slot and opening sizes.
  • Convenient for machine vises, leveling, and adjusting work on various machines.
  • Parallels slide smoothly for easy adjustment.
  • All parallels are 9/32 in (7 mm) thick.
  • Smaller sizes lock with one screw, larger sizes with two.
  • Includes a set of 6 parallels in a case.