Learn More
MSC Starrett Adjustable Parallel Set, 9/32 in Thick, 3/8 to 1-5/16 in Range, 4 Piece
Catalog No. 5000531578
Encompass
This Adjustable Parallel Set is ideal for layout, gaging, inspection, and machine tool setups. Its adjustable design allows for precise sizing with a micrometer, substituting multiple solid-type parallels. It is useful for checking slot and opening sizes, and for leveling or adjusting work in machine vises on milling, grinding, shaping, planing, and drill press machines.
  • Allows for exact size adjustment by micrometer measurement, replacing several solid-type parallels.
  • Useful as gages for checking the size of slots and openings.
  • Convenient for use in machine vises for leveling or adjusting work on various machine setups.
  • Parallels slide smoothly and are easily adjusted.
  • All parallels are 9/32 in (7 mm) thick.
Catalog No. 50-005-31578 Supplier MSC Supplier No. 86405867
This Adjustable Parallel Set is ideal for layout, gaging, inspection, and machine tool setups. Its adjustable design allows for precise sizing with a micrometer, substituting multiple solid-type parallels. It is useful for checking slot and opening sizes, and for leveling or adjusting work in machine vises on milling, grinding, shaping, planing, and drill press machines.
  • Allows for exact size adjustment by micrometer measurement, replacing several solid-type parallels.
  • Useful as gages for checking the size of slots and openings.
  • Convenient for use in machine vises for leveling or adjusting work on various machine setups.
  • Parallels slide smoothly and are easily adjusted.
  • All parallels are 9/32 in (7 mm) thick.