This universal back plunger dial indicator is designed for precise measurement of small increments. It offers 0.001 inch graduations and a 0.200 inch range, making it suitable for diverse precision tasks. Featuring an anti-magnetic design and a 1/4 inch shank, it ensures reliable performance and compatibility. The indicator includes three hardened contact points and multiple attachments, all provided in a protective case.
- Versatile for various measurement applications.
- Accurate, reliable, and simple to operate.
- Rugged design with smooth operation.
- Includes three hardened contact points.
- Shank diameter: 1/4 in (6.3 mm).
- Comes with clamp, tool post holder, snug, and hole attachment.
- Provided in a protective case.
- Anti-magnetic design.
- Dial reading: 0-100.
- Graduations: 0.001 in.
- Maximum measurement: 0.200 in.