Learn More
MSC Starrett Cable, USB to RS232, 2700 Series Indicator Compatible
Catalog No. 5000534642
Encompass
This cable is designed to connect electronic indicators to peripheral devices. It facilitates communication between a 2700 Series Wisdom Electronic Indicator and either an RS232 printer or a PC-compatible RS232 port via a USB connection.
  • Connects to RS232 printers.
  • Provides USB to RS232 (PC compatible) connectivity.
  • Compatible with 2700 series electronic indicators.
Catalog No. 50-005-34642 Supplier MSC Supplier No. 86446697
This cable is designed to connect electronic indicators to peripheral devices. It facilitates communication between a 2700 Series Wisdom Electronic Indicator and either an RS232 printer or a PC-compatible RS232 port via a USB connection.
  • Connects to RS232 printers.
  • Provides USB to RS232 (PC compatible) connectivity.
  • Compatible with 2700 series electronic indicators.